missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (Boc-aminooxy)acetic acid, 98+%

Manufacturer:  Alfa Aesar™ H5410906

 View more versions of this product

Catalog No. AAH5410906



Description

Description

Applications
(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

(Boc-aminooxy)acetic acid
5g
C7H13NO5
MFCD01632027
2-2-methylpropan-2-yl oxycarbonylamino oxyacetic acid
QBXODCKYUZNZCY-UHFFFAOYSA-N
2-[(2-methylpropan-2-yl)oxycarbonylamino]oxyacetic acid
2755974
≥98%
∽115°C (decomposition)
42989-85-5
(CH3)3COCONHOCH2CO2H
6137646
Soluble in methanol.
CC(C)(C)OC(=O)NOCC(=O)O
191.183
191.18

Chemical Identifiers

42989-85-5
191.183
QBXODCKYUZNZCY-UHFFFAOYSA-N
2755974
CC(C)(C)OC(=O)NOCC(=O)O
C7H13NO5
MFCD01632027
2-2-methylpropan-2-yl oxycarbonylamino oxyacetic acid
2-[(2-methylpropan-2-yl)oxycarbonylamino]oxyacetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents