Learn More
(Boc-aminooxy)acetic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5410903
Description
(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
(Boc-aminooxy)acetic acid | |
∽115°C (decomposition) | |
(CH3)3COCONHOCH2CO2H | |
1 g | |
boc-aminooxy acetic acid, 2-tert-butoxycarbonyl amino oxy acetic acid, boc-aoa-oh, boc-aminooxyacetic acid, boc-aoac-oh, n-boc-carboxymethoxy amine, boc-aminooxy aceticacid, 2-tert-butoxy carbonyl amino oxy acetic acid, 2-2-methylpropan-2-yl oxycarbonylamino oxyacetic acid, tert-butoxycarbonyl aminooxy acetic acid | |
QBXODCKYUZNZCY-UHFFFAOYSA-N | |
2-[(2-methylpropan-2-yl)oxycarbonylamino]oxyacetic acid | |
2755974 | |
≥98% |
42989-85-5 | |
C7H13NO5 | |
MFCD01632027 | |
6137646 | |
Soluble in methanol. | |
CC(C)(C)OC(=O)NOCC(=O)O | |
191.183 | |
191.18 |
Chemical Identifiers
42989-85-5 | |
191.183 | |
QBXODCKYUZNZCY-UHFFFAOYSA-N | |
2755974 | |
CC(C)(C)OC(=O)NOCC(=O)O |
C7H13NO5 | |
MFCD01632027 | |
boc-aminooxy acetic acid, 2-tert-butoxycarbonyl amino oxy acetic acid, boc-aoa-oh, boc-aminooxyacetic acid, boc-aoac-oh, n-boc-carboxymethoxy amine, boc-aminooxy aceticacid, 2-tert-butoxy carbonyl amino oxy acetic acid, 2-2-methylpropan-2-yl oxycarbonylamino oxyacetic acid, tert-butoxycarbonyl aminooxy acetic acid | |
2-[(2-methylpropan-2-yl)oxycarbonylamino]oxyacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only