missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-β-alanine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 437650050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
77.5°C to 78.5°C | |
3303-84-2 | |
99% | |
Authentic | |
Glass Bottle | |
WCFJUSRQHZPVKY-UHFFFAOYSA-N | |
3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
76809 | |
99% | |
Crystalline Powder |
BOC-β-Alanine | |
White | |
C8H15NO4 | |
MFCD00037291 | |
boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid | |
CC(C)(C)OC(=O)NCCC(O)=O | |
189.21 | |
189.21 | |
5 g |
Chemical Identifiers
3303-84-2 | |
189.21 | |
WCFJUSRQHZPVKY-UHFFFAOYSA-N | |
76809 | |
CC(C)(C)OC(=O)NCCC(O)=O |
C8H15NO4 | |
MFCD00037291 | |
boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid | |
3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Safety and Handling
EINECSNumber : 221-979-2
RUO – Research Use Only