Please login to your online account to display your discounted pricing

BNTX maleate, Tocris


Standard δ1 selective antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS089910

 View more versions of this product

Catalog No. 089910

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 545.59
Molecular Formula C27H27NO4·C4H4O4
Assay Percent Range >99%
Chemical Name or Material BNTX maleate
CAS 864461-31-4
Synonym 7-Benzylidenenaltrexone maleate
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material BNTX maleate
Assay Percent Range >99%
Molecular Formula C27H27NO4·C4H4O4
Formula Weight 545.59
Synonym 7-Benzylidenenaltrexone maleate
Accessories

Desiccate at -20°C