Please login to your online account to display your discounted pricing

BNTX maleate, Tocris

Standard δ1 selective antagonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material BNTX maleate
Assay Percent Range >99%
Molecular Formula C27H27NO4·C4H4O4
Formula Weight 545.59
Synonym 7-Benzylidenenaltrexone maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

089910

 
r & d systems
RDS089910
10mg Each for $214.12

089950

 
r & d systems
RDS089950
50mg Each for $895.76
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 545.59
Molecular Formula C27H27NO4·C4H4O4
Assay Percent Range >99%
Chemical Name or Material BNTX maleate
CAS 864461-31-4
Synonym 7-Benzylidenenaltrexone maleate
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C