Please login to your online account to display your discounted pricing

BMY 7378 dihydrochloride, Tocris

Selective α1D antagonist

$89.94 - $384.55

Chemical Identifiers

Chemical Name or Material BMY 7378 dihydrochloride
Assay Percent Range >98%
Molecular Formula C22H31N3O3·2HCl
Formula Weight 458.42
Synonym 8-[2-[4-(methoxyphenyl)-1-piperazinyl]ethyl]-8-azaspiro[4.5]decane-7, 9-dione dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

100610

 
r & d systems
100610
10mg Each for $89.94

100650

 
r & d systems
100650
50mg Each for $384.55
Description & Specifications

Specifications

Formula Weight 458.42
Quantity 10mg
Chemical Name or Material BMY 7378 dihydrochloride
Assay Percent Range >98%
Molecular Formula C22H31N3O3·2HCl
CAS 21102-95-4
Synonym 8-[2-[4-(methoxyphenyl)-1-piperazinyl]ethyl]-8-azaspiro[4.5]decane-7, 9-dione dihydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature