Please login to your online account to display your discounted pricing

BMY 14802 hydrochloride, Tocris

Sigma antagonist; Antipsychotic agent

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material BMY 14802 hydrochloride
Assay Percent Range >98%
Molecular Formula C18H22F2N4O·HCl
Formula Weight 384.86
Synonym α-(4-Fluorophenyl)-4-(5-fluoro-2-pyrimidinyl)-1-piperazinebutanol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

144010

 
r & d systems
144010
10mg Each for $168.15

144050

 
r & d systems
144050
50mg Each for $697.40
Description & Specifications

Specifications

Formula Weight 384.86
Quantity 10mg
Chemical Name or Material BMY 14802 hydrochloride
Assay Percent Range >98%
Molecular Formula C18H22F2N4O·HCl
CAS 105565-55-7
Synonym α-(4-Fluorophenyl)-4-(5-fluoro-2-pyrimidinyl)-1-piperazinebutanol hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature