Please login to your online account to display your discounted pricing

BMS CCR2 22, Tocris

High affinity, potent CCR2 antagonist

$153.56 - $1,362.21

Chemical Identifiers

Chemical Name or Material BMS CCR2 22
Assay Percent Range >98%
Molecular Formula C28H34F3N5O4S
Formula Weight 593.66
Synonym 2-[(Isopropylaminocarbonyl)amino]-N-[2-[[cis-2-[[4-(methylthio)benzoyl]amino]cyclohexyl]amino]-2-oxoethyl]-5-(trifluoromethyl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

31291

 
r & d systems
RDS31291
1mg Each for $153.56

312950

 
r & d systems
RDS312950
50mg Each for $1,362.21

312910

 
r & d systems
RDS312910
10mg Each for $330.06
Description & Specifications

Specifications

Formula Weight 593.66
Quantity 1mg
Chemical Name or Material BMS CCR2 22
Assay Percent Range >98%
Molecular Formula C28H34F3N5O4S
CAS 445479-97-0
Synonym 2-[(Isopropylaminocarbonyl)amino]-N-[2-[[cis-2-[[4-(methylthio)benzoyl]amino]cyclohexyl]amino]-2-oxoethyl]-5-(trifluoromethyl)benzamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C