Please login to your online account to display your discounted pricing

BMS 753, Tocris™

RARα-selective agonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material BMS 753
Assay Percent Range >99%
Molecular Formula C21H21NO4
Formula Weight 351.4
Synonym 4-[[(2,3-Dihydro-1,1,3,3-tetramethyl-2-oxo-1H-inden-5-yl)carbonyl]amino]benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

350550

 
r & d systems
RDS350550
50mg Each for $924.23

350510

 
r & d systems
RDS350510
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 351.4
Quantity 50mg
Chemical Name or Material BMS 753
Assay Percent Range >99%
Molecular Formula C21H21NO4
CAS 215307-86-1
Synonym 4-[[(2,3-Dihydro-1,1,3,3-tetramethyl-2-oxo-1H-inden-5-yl)carbonyl]amino]benzoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature