Please login to your online account to display your discounted pricing

BMS 493, Tocris™

Pan-RAR inverse agonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material BMS 493
Assay Percent Range >98%
Molecular Formula C29H24O2
Formula Weight 404.5
Synonym 4-[(1E)-2-[5, 6-Dihydro-5, 5-dimethyl-8-(2-phenylethynyl)-2-naphthalenyl]ethenyl]benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

350950

 
r & d systems
RDS350950
50mg Each for $997.23

350910

 
r & d systems
RDS350910
10mg Each for $244.81
Description & Specifications

Specifications

Formula Weight 404.5
Quantity 50mg
Assay Percent Range >98%
Chemical Name or Material BMS 493
Molecular Formula C29H24O2
CAS 215030-90-3
Synonym 4-[(1E)-2-[5, 6-Dihydro-5, 5-dimethyl-8-(2-phenylethynyl)-2-naphthalenyl]ethenyl]benzoic acid
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C