Please login to your online account to display your discounted pricing

BMS 470539 dihydrochloride, Tocris™

Potent, selective MC1 receptor agonist

$297.57 - $1,231.36

Chemical Identifiers

Chemical Name or Material BMS 470539 dihydrochloride
Assay Percent Range >98%
Molecular Formula C32H41N5O4·2HCl
Formula Weight 632.62
Synonym 1-[1-(3-Methyl-L-histidyl-O-methyl-D-tyrosyl)-4-phenyl-4-piperidinyl]-1-butanone dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

405350

 
r & d systems
RDS405350
50mg Each for $1,231.36

405310

 
r & d systems
RDS405310
10mg Each for $297.57
Description & Specifications

Specifications

Formula Weight 632.62
Quantity 50mg
Chemical Name or Material BMS 470539 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Synonym 1-[1-(3-Methyl-L-histidyl-O-methyl-D-tyrosyl)-4-phenyl-4-piperidinyl]-1-butanone dihydrochloride
CAS 457893-92-4
Molecular Formula C32H41N5O4·2HCl
Assay Percent Range >98%
Safety and Handling

Desiccate at Room Temperature