Please login to your online account to display your discounted pricing

BMS 193885, Tocris

Potent, competitive NPY Y1 receptor antagonist

$303.99 - $1,257.94

Chemical Identifiers

Chemical Name or Material BMS 193885
Assay Percent Range >99%
Molecular Formula C33H42N4O6·C3H6O3
Formula Weight 680.79
Synonym 1,4-Dihydro-4-[3-[[[[3-[4-(3-methoxyphenyl)-1-piperidinyl]propyl]amino]carbonyl]amino]phenyl]-2,6-dimethyl-3,5-pyridinedicarboxylic acid 3,5-dimethyl ester L-Lactate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

324250

 
r & d systems
RDS324250
50mg Each for $1,257.94

324210

 
r & d systems
RDS324210
10mg Each for $303.99
Description & Specifications

Specifications

Formula Weight 680.79
Quantity 50mg
Chemical Name or Material BMS 193885
Molecular Formula C33H42N4O6·C3H6O3
Assay Percent Range >99%
CAS 679839-66-8
Synonym 1,4-Dihydro-4-[3-[[[[3-[4-(3-methoxyphenyl)-1-piperidinyl]propyl]amino]carbonyl]amino]phenyl]-2,6-dimethyl-3,5-pyridinedicarboxylic acid 3,5-dimethyl ester L-Lactate
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature