missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BMS 182874 hydrochloride, Tocris

Highly selective, orally active non-peptide ETA antagonist

$250.58 - $1,042.86

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C17H19N3O3S·HCl
Formula Weight 381.88
Synonym 5-(Dimethylamino)-N-(3, 4-dimethyl-5-isoxazolyl)-1-naphthalenesulfonamide hydrochloride
Chemical Name or Material BMS 182874 hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
144110 R & D Systems
RDS144110
10mg Each for $250.58
144150 R & D Systems
RDS144150
50mg Each for $1,042.86
Specifications Description & Specifications

Chemical Identifiers

>98%
381.88
BMS 182874 hydrochloride
C17H19N3O3S·HCl
5-(Dimethylamino)-N-(3, 4-dimethyl-5-isoxazolyl)-1-naphthalenesulfonamide hydrochloride

Specifications

10mg
>98%
381.88
BMS 182874 hydrochloride
1215703-04-0
C17H19N3O3S·HCl
5-(Dimethylamino)-N-(3, 4-dimethyl-5-isoxazolyl)-1-naphthalenesulfonamide hydrochloride

Safety and Handling

Recommended Storage : Store at Room Temperature