missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ BMPR2 Recombinant Human Protein DFS Item

Recombinant human BMPR2 (174-end) was expressed using baculovirus in Sf9 insect cells using an N-terminal His tag.

Manufacturer:  Invitrogen™ LSPV6258

 View more versions of this product

Catalog No. PV6258



Description

Description

BMPR2 is a member of the bone morphogenetic protein (BMP) receptor family of transmembrane serine/threonine kinases. The ligands of this receptor are BMPs that are involved in endochondral bone formation and embryogenesis. The loss of interaction and lack of phosphorylation of TCTEL1 by BMPR2 may contribute to the pathogenesis of primary pulmonary hypertension (PPH) and BMPR2 also plays an essential role in human T-cell differentiation.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
97.1kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
aa 174-end
Yes
50mM sodium phosphate pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT, 25% glycerol
1mg
Kinase Assay
Human
His Tag
Documents