missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human BMPR1A (ALK3), GST Tag Recombinant Protein
Human BMPR1A (ALK3), GST Tag Recombinant Protein
$776.00 - $2,650.00
Specifications
Accession Number | NP_004320.2 |
---|---|
Gene Alias | BMPR1A (ALK3) |
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV6038
|
Invitrogen™
PV6038 |
10μg |
N/A
|
|
|||||
PR9136A
|
Invitrogen™
PR9136A |
100μg |
N/A
|
|
|||||
PV6039
|
Invitrogen™
PV6039 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
BMPR1A phosphorylates intracellular SMADs in response to bone morphogenic proteins.
Specifications
NP_004320.2 | |
Human | |
GST Tag |
BMPR1A (ALK3) | |
Recombinant |