Invitrogen™ BMPR1A (ALK3) Recombinant Human Protein DFS Item
BMPR1A is a Type I member of the TGF beta receptor superfamily of transmembrane ser/thr kinases.
Manufacturer:  Invitrogen™ LSPV6039
 View more versions of this product
Catalog No. PV6039
Description
Description
BMPR1A phosphorylates intracellular SMADs in response to bone morphogenic proteins.
Specifications
Specifications
NP_004320.2
Frozen
1mg
RUO - research use only
BMPR1A (ALK3)
Insect
Human
GST Tag
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
67kDa
Kinase proteins are shipped on dry ice and should be stored at -80°C.
Catalytic
Kinase Assay
STKR Kinase Family
Yes
Documents