Invitrogen™ BMPR1A (ALK3) Recombinant Human Protein DFS Item
BMPR1A is a Type I member of the TGF beta receptor superfamily of transmembrane ser/thr kinases.
Manufacturer:  Invitrogen™ LSPV6038
 View more versions of this product
Catalog No. PV6038
Description
Description
BMPR1A phosphorylates intracellular SMADs in response to bone morphogenic proteins.
Specifications
Specifications
NP_004320.2
Frozen
67kDa
Kinase proteins are shipped on dry ice and should be stored at -80°C.
Full Length, Catalytic
BMPR1A (ALK3)
Kinase Assay
Insect
aa 187-532
Yes
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
657
10μg
N-Terminal
Serine/Threonine Kinases
BMPR1A
Kinases & Inhibitors
STKR Kinase Family
Human
GST Tag
Documents