Please login to your online account to display your discounted pricing

BML-190, Tocris

Potent, selective CB2 ligand

$133.98 - $567.83

Chemical Identifiers

Chemical Name or Material BML-190
Assay Percent Range >99%
Molecular Formula C23H23ClN2O4
Formula Weight 426.9
Synonym 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1-[2-(4-morpholinyl)-2-oxoethyl]-1H-indole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

138350

 
r & d systems
RDS138350
50mg Each for $567.83

138310

 
r & d systems
RDS138310
10mg Each for $133.98
Description & Specifications

Specifications

Formula Weight 426.9
Quantity 50mg
Chemical Name or Material BML-190
Assay Percent Range >99%
CAS 2854-32-2
Molecular Formula C23H23ClN2O4
Synonym 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1-[2-(4-morpholinyl)-2-oxoethyl]-1H-indole
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature