Please login to your online account to display your discounted pricing

Thermo Scientific™ Bme1390I (ScrFI)

Cut at CC^NGG sites with Bme1390I (ScrFI) restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: BmrFI, BssKI, BstSCI, MspR9I, ScrFI, StyD4I).

$43.17 - $43.17

Specifications

Components 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100 mM NaCl and 0.1mg/mL BSA
Concentration 10U/µL
Incubator Temperature 37°C
Product Name Bme1390I (ScrFI)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

FERER1421

 
thermo scientific™
ER1421
500U Each for $43.17
Description & Specifications

Specifications

Components 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100 mM NaCl and 0.1mg/mL BSA
Concentration 10U/µL
Incubator Temperature 37°C
Product Name Bme1390I (ScrFI)

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities