Propper Blue Autoclavable Tape
Manufacturer:  Propper 26800800
 View more versions of this product
Catalog No. 11-889-4
Alert:
Cannot be used for ethylene oxide or dry heat sterilization. Do not use on linen or muslin wraps.
Description
Description
  • Specially designed with an aggressive adhesive backing to stick securely to all disposable wraps
  • High-contrast, white, steam-sensitive stripes turn dark when subjected to steam autoclaving temperatures
Order Info
In 60 yd. (54.5m) lengths
Specifications
Specifications
Autoclavable
Synthetic resin
Tape
Indicator Tape
1 Ea.
54.9 m
3 in.
Rubber
Blue,White
Crepe Paper
Different types of sterilization packs, to distinguish processed and unprocessed items
180 ft.
0.75 in.
7.62 cm