Please login to your online account to display your discounted pricing

Blood Group H1 (O) Type 1 Mouse anti-Human, Clone: 17-206, eBioscience™

Mouse Monoclonal Antibody

$131.95 - $323.35

Specifications

Antigen Blood Group H1 (O) Type 1
Clone 17-206
Host Species Murine
Species Reactivity Human
Applications ELISA
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122875

 
life technologies
LS14981080
25μg Each for $131.95

501122876

 
life technologies
LS14981082
100μg Each for $323.35
Description & Specifications

Specifications

Antigen Blood Group H1 (O) Type 1
Clone 17-206
Host Species Murine
Species Reactivity Human
Applications ELISA
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 17-206 recognizes the human blood group antigen H (O) type 1, a subtype primarily found in epithelial and secretory cells. The precursor H antigen is modified by addition of saccharides to carbohydrate chains of proteins and lipids on erythrocytes and epithelial cells. The type, attachment, location, and sequence of saccharides determines the blood group antigen subtype and is mediated by multiple glycosyl transferases. Depending on ABO blood type, the H antigen is converted into either the A antigen, B antigen, both the A and B antigen, or left unconverted (O antigen). Evidence that the blood group antigen H is not expressed in tumor cells and that the level correlates with disease progression suggests that the blood group antigen H may be involved in cell proliferation, cell adhesion, and angiogenesis, although its exact function remains to be determined. The 17-206 antibody does not recognize blood group antigen H (O) type 2.