Learn More
Thermo Scientific™ Blocker™ Casein
Block membranes, plates and other samples with these ready-to-use PBS or TBS solutions of purified casein for Western blot, ELISA and other detection methods.
$72.92 - $280.87
Specifications
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
PI37528
|
Thermo Scientific™
PI37528 |
Blocker Casein in PBS | 1 L |
Pack for $259.02
|
|
PI37532
|
Thermo Scientific™
PI37532 |
Blocker Casein in TBS | 1 L |
Pack for $280.87
|
|
PI37582
|
Thermo Scientific™
37582 |
Blocker Casein in PBS | 100 mL |
Each for $75.06
|
|
PI37583
|
Thermo Scientific™
37583 |
Blocker Casein in TBS | 100 mL |
Each for $72.92
|
|
Description
Thermo Scientific™ Blocker Casein Blocking Buffers are ready-to-use, PBS or TBS solutions of purified casein protein for blocking steps in Western blotting, ELISA, immunohistochemistry and nucleic acid detection methods.
These blocking buffers contains casein protein that is purified from milk by the Hammarsten method. The phosphate- and Tris-buffered saline formulations are used for blocking excess binding sites on membranes and microplates in antibody-based detection methods. Blocker Casein Buffers are 1% (w/v) casein, which corresponds to the optimal concentration for most applications.Highlights:
Purified casein – single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
Easy to use – 1% casein solutions are ready to use; can be diluted further as needed
Flexible – available in PBS and TBS formulations to suit a variety of applications
Safe – stable, thimerosal-free formulations
Specifications
0.0 | |
icon-pdf.png |