Learn More
Blimp-1 Monoclonal Antibody (5E7), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Supplier: Invitrogen 12985082
Description
Description: This 5E7 monoclonal antibody recognizes mouse B lymphocyte-induced maturation protein-1 (Blimp-1), which is also known as Prdm1. Blimp-1 is a transcriptional repressor with five zinc finger domains. Blimp-1 is expressed by terminally differentiating B and T cells. Blimp-1 is also expressed by some dendritic cells, and some NK cells. This transcriptional repressor is required for B cell differentiation into plasma cells and serves as a critical antagonist of CD4+ follicular helper T cell differentiation. While Blimp-1 is not required for priming and development of T and B cell effector function, it is critical for effector cell terminal differentiation. Blimp-1 promotes effector molecule secretion while suppressing proliferation. Blimp-1 also acts as a transcriptional antagonist of Bcl-6 and inhibits genes that are related to memory cell differentiation. Applications Reported: The 5E7 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: The 5E7 antibody has been tested by flow cytometric analysis of splenocytes isolated from sheep red blood cell-immunized mice. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Blimp-1 (B lymphocyte maturation protein-1) is a nuclear protein with five zinc fingers in its carboxy terminus. It is a transcriptional repressor that is both required and sufficient to trigger terminal differentiation of B lymphocytes and monocyte/macrophages. Blimp-1 is expressed in, and may be required for terminally differentiated epithelial cells, retinal neurons and other lineage cells. It has also been implicated in the repression of beta-interferon (IFN-beta) and c-myc gene expression.Specifications
Blimp-1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q60636 | |
PRDM1 | |
Affinity chromatography | |
RUO | |
12142 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
5E7 | |
PE | |
PRDM1 | |
B lymphocyte-induced maturation protein 1; b2b1765Clo; Beta-interferon gene positive regulatory domain I-binding factor; beta-interferon gene positive-regulatory domain I binding factor; BLIMP 1; Blimp1; BLIMP-1; B-lymphocyte-induced maturation protein 1; MGC118922; MGC118923; MGC118924; MGC118925; OTTHUMP00000016918; OTTHUMP00000202173; Positive regulatory domain I-binding factor 1; PR domain 1; PR domain containing 1, with ZNF domain; PR domain zinc finger protein 1; PR domain-containing protein 1; PR/SET domain 1; PRDI BF1; PRDI-BF1; PRDI-binding factor-1; Prdm1; RP1-134E15.1; ZNFPR1A1 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.