Please login to your online account to display your discounted pricing

Alfa Aesar™ Bleomycin sulfate, Cell Culture Grade


Quantity 15mg

Manufacturer: alfa aesar™  J67560SDL

 View more versions of this product

Catalog No. AAJ67560SDL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

MDL Number MFCD00070310
Chemical Name or Material Bleomycin sulfate
CAS 9041-93-4
Health Hazard 2 GHS H Statement
H340-H351
May cause genetic defects.
Suspected of causing cancer.
Physical Form Powder
Purity Grade Cell Culture Grade
RTECS Number EC5991990
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 15mg
Health Hazard 1 Danger
Merck Index 14,1318
Solubility Information Soluble in water or DMSO
Color White

Specifications

Chemical Name or Material Bleomycin sulfate
CAS 9041-93-4
Purity Grade Cell Culture Grade
MDL Number MFCD00070310
Merck Index 14,1318

Solubility
Soluble in water or DMSO

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.

Accessories

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H340-H351
May cause genetic defects.
Suspected of causing cancer.

SDS & Safety Documents