Please login to your online account to display your discounted pricing

Alfa Aesar™ Bleomycin sulfate, Cell Culture Grade

Chemical Identifiers

Chemical Name or Material Bleomycin sulfate
CAS 9041-93-4
Purity Grade Cell Culture Grade
MDL Number MFCD00070310
Merck Index 14,1318
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67560SDL

 
alfa aesar™
J67560SDL
15mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Bleomycin sulfate
MDL Number MFCD00070310
CAS 9041-93-4
Purity Grade Cell Culture Grade
Physical Form Powder
Merck Index 14,1318
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H340-H351
May cause genetic defects.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 15mg
RTECS Number EC5991990
Solubility Information Soluble in water or DMSO
Color White

Solubility
Soluble in water or DMSO

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.

Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H340-H351
May cause genetic defects.
Suspected of causing cancer.