Please login to your online account to display your discounted pricing
Black Stallion™ Jumbo Cover Mitts
$45.34 - $139.89
Products
Catalog Number Mfr. No. Price Quantity    
19-056609
revco industries
HPC3
Pair for $139.89
19-057795
revco industries
DKC1R
Pair for $45.34
Description & Specifications
Designed to provide added wear, protection, and insulation when worn over gloves and mitts. Reversible 22 oz. (624g) Kevlar™ style provides protection at temperatures up to 399°C (750°F).
Sold by the pair.