Please login to your online account to display your discounted pricing
Black Stallion™ High-Temperature Double-Lined Gloves
$74.27 - $133.01
Specifications
Length (English) 14 in.
Length (Metric) 35.5cm
Size One Size
For Use With (Application) foundry work, Metal curing and refining, Aluminum extrusion and welding
View More Specs
Products
Catalog Number Mfr. No. Temperature (English) Max. Weight (English) Material Price Quantity    
19-056530
revco industries
P114
1400°F 22 oz. PBI™ Pair for $133.01
19-057180
revco industries
G114
1000°F 36 oz. Fiberglass Pair for $74.30
19-057282
revco industries
DK114
750°F 22 oz. Kevlar™ Pair for $74.27
Description & Specifications
Specifications
Length (English) 14 in.
Length (Metric) 35.5cm
Size One Size
For Use With (Application) foundry work, Metal curing and refining, Aluminum extrusion and welding
Heat-resistant gloves in three materials, each with double-lined palm for superior insulation. Ideal for foundry work, metal curing and refining, aluminum extrusion, and welding.
All three styles measure 14 in. long (36 cm). Sold by the pair.