Fisherbrand™ Black Ribbed Phenolic Cap with PE Cone Liner
Forms exceptionally tight seal, offers good chemical barrier
Manufacturer:  Fisherbrand™ ALP276940L
 View more versions of this product
Catalog No. 02-912-058A
Description
Description
  • Form tight seal
  • Offers good chemical barrier
  • Used to seal liquids, soaps, macro-invertebrates
Specifications
Specifications
Phenolic
Black
PE
Polycone cap
Black Ribbed
15-425
Polycone
Sealing
12/Pk.
Yes
Polycone
Screw
missing translation for 'documents'