Please login to your online account to display your discounted pricing

Qorpak™ Black Phenolic Pulp/Tinfoil Lined Caps

For use with solids and powders. Qorpak™ Black Phenolic Pulp/Tinfoil Lined Caps offer strong resistance to chemicals and temperature.

$1,312.27 - $1,312.27

Specifications

Closure Liner Pulp/Tin Foil
Color Black
View More Specs
Products
Catalog Number Mfr. No. Closure Type Neck Finish No. per Case Packaging Price Quantity    

02-993-353

 
qorpak™
CAP00268
Smooth 48-400 576 Cs. Bagged Case for $1,312.27
Description & Specifications

Specifications

Closure Liner Pulp/Tin Foil
Color Black

  • Not recommended for acids and alkalis
  • Available as ribbed and smooth style
  • Thread finish for narrow bottles is 425 GPI and for wider 400
  • Available as bulk/bagged packaging