Please login to your online account to display your discounted pricing

BL 1249, Tocris™

Putative K2P2.1 (TREK-1) channel opener

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material BL 1249
Assay Percent Range >99%
Molecular Formula C17H17N5
Formula Weight 291.35
Synonym 5,6,7,8-Tetrahydro-N-[2-(2H-tetrazol-5-yl)phenyl]-1-naphthalenamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

379750

 
r & d systems
379750
50mg Each for $904.70

379710

 
r & d systems
379710
10mg Each for $221.01
Description & Specifications

Specifications

Formula Weight 291.35
Quantity 50mg
Assay Percent Range >99%
Molecular Formula C17H17N5
Synonym 5,6,7,8-Tetrahydro-N-[2-(2H-tetrazol-5-yl)phenyl]-1-naphthalenamine
Recommended Storage Store at -20°C
Chemical Name or Material BL 1249
CAS 18200-13-0
Safety and Handling

Store at -20°C