Please login to your online account to display your discounted pricing

BIX 01294, Tocris

G9a-like protein and G9a histone lysine methyltransferase inhibitor

$238.81 - $984.19

Chemical Identifiers

Chemical Name or Material BIX 01294
Assay Percent Range >98%
Molecular Formula C28H38N6O2·3HCl
Formula Weight 600.02
Synonym 2-(Hexahydro-4-methyl-1H-1, 4-diazepin-1-yl)-6, 7-dimethoxy-N-[1-(phenylmethyl)-4-piperidinyl]-4-quinazolinamine trihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

336410

 
r & d systems
336410
10mg Each for $238.81

336450

 
r & d systems
336450
50mg Each for $984.19
Description & Specifications

Specifications

Formula Weight 600.02
Quantity 10mg
Chemical Name or Material BIX 01294
Assay Percent Range >98%
Molecular Formula C28H38N6O2·3HCl
CAS 1392399-03-9
Synonym 2-(Hexahydro-4-methyl-1H-1, 4-diazepin-1-yl)-6, 7-dimethoxy-N-[1-(phenylmethyl)-4-piperidinyl]-4-quinazolinamine trihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature