Please login to your online account to display your discounted pricing

Biuret Reagent TS, Ricca Chemical


Quantity 1L
Packaging Poly natural

Manufacturer: ricca chemical company  RI102532

 View more versions of this product

Catalog No. 1025-32

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Clear blue
Physical Form Liquid
Quantity 1L
CAS Max % 95.0
CAS Min % 91.0
CAS 7732-18-5
Packaging Poly natural
CAS 6381-59-5
CAS Max % 0.52
CAS 7758-99-8
CAS Min % 0.13
CAS Max % 0.13
CAS Min % 6.22
CAS Max % 6.48
CAS Min % 0.5
Chemical Name or Material Biuret Reagent
Name Note Test Solution
CAS 1310-73-2

Specifications

Chemical Name or Material Biuret Reagent
Name Note Test Solution
CAS 7732-18-5
CAS Min % 91.0
CAS Max % 95.0
CAS 1310-73-2
CAS Min % 6.22
CAS Max % 6.48
CAS 6381-59-5
CAS Min % 0.5
CAS Max % 0.52
CAS 7758-99-8
CAS Min % 0.13
CAS Max % 0.13
Physical Form Liquid