missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bisphenol F acetate propionate, epoxy curative, EC-298

Well suited for use in epoxy/(meth)acrylate hybrid thermoset compositions

Manufacturer:  Alfa Aesar™ 4672422

 View more versions of this product

Catalog No. AA4672422

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Bisphenol F acetate propionate
1071466-53-9
MFCD22380705
JARUUDOWTQBKKU-UHFFFAOYSA-N
[4-[(4-acetyloxyphenyl)methyl]phenyl] propanoate
59317436
Liquid
100g
Epoxy curative, EC-298
C18H18O4
4-4-acetyloxy phenyl methyl phenyl propanoate, bisphenol f acetate propionate, epoxy curative, ec-298
CCC(=O)OC1=CC=C(C=C1)CC2=CC=C(C=C2)OC(=O)C
298.338
298.34
Yellow
150°C (302°F)

Chemical Identifiers

1071466-53-9
298.338
JARUUDOWTQBKKU-UHFFFAOYSA-N
59317436
CCC(=O)OC1=CC=C(C=C1)CC2=CC=C(C=C2)OC(=O)C
C18H18O4
MFCD22380705
4-4-acetyloxy phenyl methyl phenyl propanoate, bisphenol f acetate propionate, epoxy curative, ec-298
[4-[(4-acetyloxyphenyl)methyl]phenyl] propanoate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2°C to 8°C

SDS Documents