Please login to your online account to display your discounted pricing

Bisphenol A diacetate, 99+%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  249311000

 View more versions of this product

Catalog No. AC249311000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 312.34
Quantity 100g
Color White to off-white
EINECS Number 233-472-3
TSCA TSCA
Melting Point 86° to 91°C
Chemical Name or Material Bisphenol A diacetate
Assay Percent Range 99+%
Molecular Formula C19H20O4
Linear Formula {CH3CO2C6H4}2C(CH3)2
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Name Note 99+%
MDL Number MFCD00026194
CAS 10192-62-8
Synonym 2, 2-Bis-(4-hydroxyphenol)propane diacetate

Specifications

Chemical Name or Material Bisphenol A diacetate
Name Note 99+%
CAS 10192-62-8
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C19H20O4
Linear Formula {CH3CO2C6H4}2C(CH3)2
Formula Weight 312.34
MDL Number MFCD00026194
Synonym 2, 2-Bis-(4-hydroxyphenol)propane diacetate
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents