missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bisphenol A acetate propionate, epoxy curative, EC-326

Manufacturer:  Alfa Aesar™ 4644122

 View more versions of this product

Catalog No. AA4644122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Recommended for use as an epoxy curative to improve toughness, hydrophobicity, and hydrolytic stability
Specifications

Specifications

Bisphenol A acetate propionate
1071466-57-3
MFCD22380701
VWMUCEDXVAQLNS-UHFFFAOYSA-N
[4-[2-(4-acetyloxyphenyl)propan-2-yl]phenyl] propanoate
59317407
Solid
100g
Epoxy curative, EC-326
C20H22O4
bisphenol a acetate propionate, technical grade, bisphenol a acetate propionate, epoxy curative, ec-326, 4-2-4-acetyloxy phenyl propan-2-yl phenyl propanoate
CCC(=O)OC1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)OC(=O)C
326.392
326.39
White to Yellow

Chemical Identifiers

1071466-57-3
326.392
VWMUCEDXVAQLNS-UHFFFAOYSA-N
59317407
CCC(=O)OC1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)OC(=O)C
C20H22O4
MFCD22380701
bisphenol a acetate propionate, technical grade, bisphenol a acetate propionate, epoxy curative, ec-326, 4-2-4-acetyloxy phenyl propan-2-yl phenyl propanoate
[4-[2-(4-acetyloxyphenyl)propan-2-yl]phenyl] propanoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2°C to 8°C

SDS Documents