Please login to your online account to display your discounted pricing

(±)-Bisoprolol hemifumarate, Tocris

β1 antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material (±)-Bisoprolol hemifumarate
Assay Percent Range >98%
Molecular Formula C18H31NO4·1/2C4H4O4
Formula Weight 383.48
Synonym (±)-1-[4-[[2-(1-Methylethoxy)ethoxy)methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol hemifumarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

091410

 
r & d systems
RDS091410
10mg Each for $201.36

091450

 
r & d systems
RDS091450
50mg Each for $844.73
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 383.48
Molecular Formula C18H31NO4·1/2C4H4O4
Assay Percent Range >98%
CAS 104344-23-2
Chemical Name or Material (±)-Bisoprolol hemifumarate
Synonym (±)-1-[4-[[2-(1-Methylethoxy)ethoxy)methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol hemifumarate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature