Please login to your online account to display your discounted pricing

(±)-Bisoprolol hemifumarate, Tocris


β1 antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS091450

 View more versions of this product

Catalog No. 091450

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 383.48
CAS 104344-23-2
Chemical Name or Material (±)-Bisoprolol hemifumarate
Recommended Storage Store at Room Temperature
Synonym (±)-1-[4-[[2-(1-Methylethoxy)ethoxy)methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol hemifumarate
Molecular Formula C18H31NO4·1/2C4H4O4
Assay Percent Range >98%

Specifications

Chemical Name or Material (±)-Bisoprolol hemifumarate
Assay Percent Range >98%
Molecular Formula C18H31NO4·1/2C4H4O4
Formula Weight 383.48
Synonym (±)-1-[4-[[2-(1-Methylethoxy)ethoxy)methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol hemifumarate
Accessories

Store at Room Temperature