Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth titanium isopropoxide, 5% w/v in isopropanol

Preparation of thin film mixed oxides of Ti and Bi

Chemical Identifiers

Chemical Name or Material Bismuth titanium isopropoxide
CAS 338391-61-0
Molecular Formula C21H49BiO7Ti
Linear Formula BiTi[OCH(CH3)2]7
Formula Weight 670.48
MDL Number MFCD01865878
UN Number UN1219
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42839AC

  SDS
alfa aesar™
42839AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 670.48
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Concentration 5% w/v in isopropanol
Health Hazard 1 Danger
MDL Number MFCD01865878
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Pale yellow
Linear Formula BiTi[OCH(CH3)2]7
Chemical Name or Material Bismuth titanium isopropoxide
Quantity 25mL
CAS 338391-61-0
Sensitivity Moisture Sensitive
UN Number UN1219
Health Hazard 2 GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Storage Note 1
  • Packaged under Argon
Molecular Formula C21H49BiO7Ti
Physical Form Liquid

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

  • Packaged under Argon