Learn More
Bismuth subsalicylate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04039530
Description
To impart pearly surface to cellulose-base, polystyrene, and phenolformaldehyde resins. Typical tap bulk density 0.43g/cm^3Bismuth subsalicylate is used to treat temporary discomforts of the stomach and gastrointestinal tract, such as diarrhea, indigestion, heartburn and nausea. It also acts as an antacid. It is practiced to decrease the number of bowel movements and to make the stool firmer. It is also used to reduce inflammation within the intestine. It is further used in growth media for selective isolation of microorganisms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth subsalicylate | |
14882-18-9 | |
250 g | |
C7H6BiO4 | |
bismuth subsalicylate, bismuth oxysalicylate, wismutsubsalicylat, bismuth oxide salicylate, stabisol, vismut, bismuthi subsalicylas, spiromak forte, pepto-bismol, bismutum subsalicylicum | |
QBWLKDFBINPHFT-UHFFFAOYSA-L | |
1,3,2$l^{2}-benzodioxabismin-4-one;hydrate | |
16682734 | |
Odorless |
White | |
Powder | |
96% | |
MFCD00085368 | |
Practically insoluble in water,alcoholSoluble in acid and alkali. Insoluble in cold water,ethanol,diethyl ether and n-octanol. | |
C1=CC=C2C(=C1)C(=O)O[Bi]O2.O | |
363.101 | |
362.09 | |
White powder |
Chemical Identifiers
14882-18-9 | |
363.101 | |
QBWLKDFBINPHFT-UHFFFAOYSA-L | |
16682734 | |
C1=CC=C2C(=C1)C(=O)O[Bi]O2.O |
C7H6BiO4 | |
MFCD00085368 | |
bismuth subsalicylate, bismuth oxysalicylate, wismutsubsalicylat, bismuth oxide salicylate, stabisol, vismut, bismuthi subsalicylas, spiromak forte, pepto-bismol, bismutum subsalicylicum | |
1,3,2$l^{2}-benzodioxabismin-4-one;hydrate |
Safety and Handling
H315-H319-H335
EINECSNumber : 238-953-1
RTECSNumber : EB2985000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only