Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth strontium calcium copper oxide, (2-2-1-2), 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Bismuth strontium calcium copper oxide, (2-2-1-2)
CAS 114901-61-0
MDL Number MFCD00213932
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4046709

 
alfa aesar™
4046709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 10g
Quantity 10g
Chemical Name or Material Bismuth strontium calcium copper oxide, (2-2-1-2)
CAS 114901-61-0
Physical Form Fine Agglomerate
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00213932
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.