missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth Standard For ICP, 1000μg/mL in 10% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$196.69 - $342.51
Chemical Identifiers
CAS | 7440-69-9 |
---|---|
Molecular Formula | Bi |
Molecular Weight (g/mol) | 208.98 |
InChI Key | JCXGWMGPZLAOME-UHFFFAOYSA-N |
PubChem CID | 5359367 |
ChEBI | CHEBI:33301 |
IUPAC Name | bismuth |
SMILES | [Bi] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Bi | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-69-9 | |
208.98 | |
5359367 | |
bismuth |
Bi | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
CHEBI:33301 | |
[Bi] |
Specifications
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation | |
0.1,10.0,89.9 | |
Bi | |
[Bi] | |
208.98 | |
CHEBI:33301 | |
Triple-leached LDPE bottle | |
10% HNO3 | |
Bismuth Standard |
Matrix Concentration Cat. No. 10% HNO3 1000μg/mL PLBI4-2Y, -2X |
|
7440-69-9,7697-37-2,7732-18-5 | |
0.1,10.0,89.9 | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
bismuth | |
5359367 | |
Assurance | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
125mL |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only