missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth rod, Puratronic™, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 47229KJ

 View more versions of this product

Catalog No. AA47229KJ



Specifications

Specifications

Bismuth rod
5 pc.
7440-69-9
Bi
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
Odorless
bismuth
5359367
99.9999%
≈7.5g/cm
10mm (0.39 in.) dia. x 145mm (5.71 in.) long
(metals basis)
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
[Bi]
208.98
CHEBI:33301
Puratronic™

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
CHEBI:33301
[Bi]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents