missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
$105.95 - $351.03
Chemical Identifiers
CAS | 7440-69-9 |
---|---|
Molecular Formula | Bi |
Molecular Weight (g/mol) | 208.98 |
MDL Number | MFCD00134033 |
InChI Key | JCXGWMGPZLAOME-UHFFFAOYSA-N |
Synonym | bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod |
PubChem CID | 5359367 |
ChEBI | CHEBI:33301 |
IUPAC Name | bismuth |
SMILES | [Bi] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-69-9 | |
208.98 | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
5359367 | |
bismuth |
Bi | |
MFCD00134033 | |
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod | |
CHEBI:33301 | |
[Bi] |
Specifications
7440-69-9 | |
MFCD00134033 | |
100 g | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
bismuth | |
5359367 | |
99.5% | |
Odorless | |
Powder | |
(metals basis) |
Bi | |
UN3089 | |
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod | |
[Bi] | |
208.98 | |
CHEBI:33301 | |
−325 Mesh | |
Apparent density: 3.2g/cm3 to 4.0g/cm3 | |
Bismuth powder |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 231-177-4
RTECSNumber : EB2600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only