missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth Powder, -200 mesh, 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1076606

 View more versions of this product

Catalog No. AA1076606



Specifications

Specifications

Bismuth
7440-69-9
9.79g/cm3
Bi
atom
[Bi]
Odorless
5359367
Powder
5g
Gray powder
99.999% (Metals basis)
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
208.98
CHEBI:33301
Gray

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
atom
CHEBI:33301
[Bi]
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

H228-H303; Flammable solid. May be harmful if swallowed.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-177-4

Documents