Please login to your online account to display your discounted pricing

Bismuth Powder, -200 mesh, 99.999% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Bismuth
CAS 7440-69-9
Physical Form Powder
Assay Percent Range 99.999% (Metals basis)
MDL Number MFCD00134033
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1076614

  SDS
alfa aesar
1076614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1076606

  SDS
alfa aesar
1076606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1076622

  SDS
alfa aesar
1076622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25g
Color Gray
DOT Information DOT Class: 4.1, Packaging Group: III
EINECS Number 231-177-4
Physical Form Powder
CAS 7440-69-9
MDL Number MFCD00134033
Appearance Gray powder
Health Hazard 2 Take precautionary measures against static discharges.
Chemical Name or Material Bismuth
Assay Percent Range 99.999% (Metals basis)
Health Hazard 1 H228-H303; Flammable solid. May be harmful if swallowed.
Odor Odorless
Safety and Handling

H228-H303; Flammable solid. May be harmful if swallowed.

Take precautionary measures against static discharges.