missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth Powder, -200 mesh, 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1076622

 View more versions of this product

Catalog No. AA1076622



Specifications

Specifications

Bismuth
7440-69-9
99.999% (Metals basis)
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
208.98
CHEBI:33301
Gray
100g
Gray powder
Bi
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
[Bi]
Odorless
5359367
Powder

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
CHEBI:33301
[Bi]
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

H228-H303; Flammable solid. May be harmful if swallowed.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-177-4

Documents