missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth Powder, -200 mesh, 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1076614

 View more versions of this product

Catalog No. AA1076614



Specifications

Specifications

Bismuth
7440-69-9
99.999% (Metals basis)
MFCD00134033
atom, bismuth, elemental, iii ion, needles, pieces, powder, rod, rod, 12.7mm 0.5in dia, shot, elongated, unii-u015tt5i8h
[Bi]
Odorless
5359367
Powder
25g
Gray powder
Bi
-200 Mesh
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
208.98
CHEBI:33301
Gray

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
atom, bismuth, elemental, iii ion, needles, pieces, powder, rod, rod, 12.7mm 0.5in dia, shot, elongated, unii-u015tt5i8h
CHEBI:33301
[Bi]
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : DOT Class: 4.1; Packaging Group: II

EINECSNumber : 231-177-4

RTECSNumber : EB2600000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents