missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth powder, -100 mesh, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 10110A3

 View more versions of this product

Catalog No. AA10110A3



Specifications

Specifications

Bismuth powder
2kg
(metals basis)
MFCD00134033
−100 Mesh
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
208.98
CHEBI:33301
99.5%
Apparent density: 3.8g/cm3 to 4.6g/cm3
7440-69-9
Bi
UN3089
atom, bismuth, elemental, iii ion, needles, pieces, powder, rod, rod, 12.7mm 0.5in dia, shot, elongated, unii-u015tt5i8h
[Bi]
Odorless
5359367
Powder, Apparent density 3.8-4.6g/cm

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
atom, bismuth, elemental, iii ion, needles, pieces, powder, rod, rod, 12.7mm 0.5in dia, shot, elongated, unii-u015tt5i8h
CHEBI:33301
[Bi]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 231-177-4

RTECSNumber : EB2600000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents