missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth polycrystalline lump, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1444236

 View more versions of this product

Catalog No. AA1444236



Specifications

Specifications

Bismuth polycrystalline lump
7440-69-9
Bi
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
[Bi]
Odorless
5359367
Lumps
Puratronic™
500g
(metals basis)
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
208.98
CHEBI:33301
99.999%

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
CHEBI:33301
[Bi]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents