missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth plasma standard solution, Alfa Aesar™, Specpure™
Manufacturer: Alfa Aesar 13822AE
Specifications
Bismuth | |
Clear liquid | |
MFCD00134033 | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
bismuth | |
5359367 | |
Liquid | |
Colorless |
7440-69-9,7732-18-5,7732-18-5 | |
Bi | |
Nitric acid: 5.0%; Bismuth: 0.1%; Water: 94.9% | |
[Bi] | |
208.98 | |
CHEBI:33301 | |
23 hPa (17mm Hg) at 20°C | |
100mL |
Chemical Identifiers
7440-69-9 | |
208.98 | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
CHEBI:33301 | |
[Bi] |
Bi | |
MFCD00134033 | |
5359367 | |
bismuth |
Safety and Handling
H314: Causes severe skin burns and eye damage.
DOTInformation : Hazard Class: 8, Packing Group: III
EINECSNumber : 231-714-2